missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Phthalyl-L-tryptophan, TCI America™

Manufacturer:  TCI America P2023200MG

 View more versions of this product

Catalog No. P2023200MG



Specifications

Specifications

N-Phthalyl-L-tryptophan
181°C
48208-26-0
C19H14N2O4
RG 108
C1=CC=C2C(=C1)C(=O)N(C2=O)C(CC3=CNC4=CC=CC=C43)C(=O)O
334.331
334.33
Yellow
200mg
≥98.0% (HPLC,T)
MFCD08705332
HPTXLHAHLXOAKV-MRXNPFEDSA-N
(2R)-2-(1,3-dioxoisoindol-2-yl)-3-(1H-indol-3-yl)propanoic acid
702560
Crystal-Powder at 20°C

Chemical Identifiers

48208-26-0
334.331
HPTXLHAHLXOAKV-MRXNPFEDSA-N
702560
C1=CC=C2C(=C1)C(=O)N(C2=O)C(CC3=CNC4=CC=CC=C43)C(=O)O
C19H14N2O4
MFCD08705332
RG 108
(2R)-2-(1,3-dioxoisoindol-2-yl)-3-(1H-indol-3-yl)propanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents