Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Phthaloyl-L-phenylalanine, 98%

Chemical Identifiers

Chemical Name or Material N-Phthaloyl-L-phenylalanine
CAS 5123-55-7
Formula Weight 295.29
MDL Number MFCD00069738
Beilstein 90080
Synonym N-Pht-Phe-OH; (S)-2-(1,3-Dioxo-2-isoindolinyl)-3-phenylpropionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6502506

  SDS
alfa aesar™
H6502506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6502514

  SDS
alfa aesar™
H6502514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 181° to 185°C
Formula Weight 295.29
MDL Number MFCD00069738
Assay 98%
CAS 5123-55-7
Chemical Name or Material N-Phthaloyl-L-phenylalanine
Physical Form Powder
Sensitivity Hygroscopic
Synonym N-Pht-Phe-OH; (S)-2-(1,3-Dioxo-2-isoindolinyl)-3-phenylpropionic acid
Quantity 5g
Color White
Storage Note 1 2° to 8°C
Beilstein 90080

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol. (almost transparency)

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Safety and Handling

2° to 8°C