Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Phthaloyl-L-glycine N-succinimidyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Phthaloyl-L-glycine N-succinimidyl ester
CAS 3397-29-3
Formula Weight 302.24
MDL Number MFCD00190994
Beilstein 1506140
Synonym Pht-Gly-Osu; N-Phthaloyl-L-glycine N-hydroxysuccinimide ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6553703

 
alfa aesar™
H6553703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6553706

 
alfa aesar™
H6553706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 302.24
Chemical Name or Material N-Phthaloyl-L-glycine N-succinimidyl ester
Physical Form Solid
Assay 95%
MDL Number MFCD00190994
CAS 3397-29-3
Beilstein 1506140
Synonym Pht-Gly-Osu; N-Phthaloyl-L-glycine N-hydroxysuccinimide ester
Quantity 1g
Sensitivity Moisture Sensitive