missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Phthaloyl-DL-glutamic Anhydride, TCI America™

Manufacturer:  TCI America P17895G

 View more versions of this product

Catalog No. P17895G



Specifications

Specifications

N-Phthaloyl-DL-glutamic Anhydride
205°C
3343-28-0
MFCD00005886
ICDLEMPZXFCQEB-UHFFFAOYSA-N
2-(2,6-dioxooxan-3-yl)isoindole-1,3-dione
92193
Crystal-Powder at 20°C
White
5g
C13H9NO5
2-Phthalimidoglutaric Anhydride
C1CC(=O)OC(=O)C1N2C(=O)C3=CC=CC=C3C2=O
259.217
259.22
≥98.0% (T)

Chemical Identifiers

3343-28-0
259.217
ICDLEMPZXFCQEB-UHFFFAOYSA-N
92193
C1CC(=O)OC(=O)C1N2C(=O)C3=CC=CC=C3C2=O
C13H9NO5
MFCD00005886
2-Phthalimidoglutaric Anhydride
2-(2,6-dioxooxan-3-yl)isoindole-1,3-dione
Safety and Handling

Safety and Handling

RTECSNumber : MA3900000

TSCA : Yes

Documents