Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(Phenylseleno)phthalimide, tech. 85%

Chemical Identifiers

Chemical Name or Material N-(Phenylseleno)phthalimide
CAS 71098-88-9
Purity Grade Technical Grade
Molecular Formula C14H9NO2Se
Formula Weight 302.19g/mol
MDL Number MFCD00010406
UN Number UN3283
Beilstein 1468701
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2112503

 
alfa aesar™
B21125-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2112506

 
alfa aesar™
B21125-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point ≈180°C
Quantity 1g
Chemical Name or Material N-(Phenylseleno)phthalimide
CAS 71098-88-9
Purity Grade Technical Grade
Assay 85%
Molecular Formula C14H9NO2Se
Formula Weight 302.19g/mol
MDL Number MFCD00010406
UN Number UN3283
Beilstein 1468701
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

−30° to −10°C