Learn More
N-Phenylrhodanine, 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 391390050
Specifications
N-Phenylrhodanine | |
97.0 | |
Authentic | |
Glass bottle | |
MFCD00100196 | |
3-phenylrhodanine, n-phenylrhodanine, 3-phenylrhodanin, rhodanine, 3-phenyl, 4-thiazolidinone, 3-phenyl-2-thioxo, 3-phenylrodanin, n-phenyl-rhodanine, 3-phenyl-2-thioxo-1,3-thiazolidin-4-one, 3-phenyl-2-thioxo-thiazolidin-4-one, 3-phenyl-rhodanin german | |
C1C(=O)N(C(=S)S1)C2=CC=CC=C2 | |
209.29 | |
209.29 |
1457-46-1 | |
100.0 | |
97% min. (HPLC) | |
C9H7NOS2 | |
5g | |
DVRWEKGUWZINTQ-UHFFFAOYSA-N | |
3-phenyl-2-sulfanylidene-1,3-thiazolidin-4-one | |
73822 | |
97+% |
Chemical Identifiers
1457-46-1 | |
209.29 | |
DVRWEKGUWZINTQ-UHFFFAOYSA-N | |
73822 | |
C1C(=O)N(C(=S)S1)C2=CC=CC=C2 |
C9H7NOS2 | |
MFCD00100196 | |
3-phenylrhodanine, n-phenylrhodanine, 3-phenylrhodanin, rhodanine, 3-phenyl, 4-thiazolidinone, 3-phenyl-2-thioxo, 3-phenylrodanin, n-phenyl-rhodanine, 3-phenyl-2-thioxo-1,3-thiazolidin-4-one, 3-phenyl-2-thioxo-thiazolidin-4-one, 3-phenyl-rhodanin german | |
3-phenyl-2-sulfanylidene-1,3-thiazolidin-4-one |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 215-940-9
RUO â Research Use Only