missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Phenylmaleimide, 98+%

Manufacturer:  Alfa Aesar™ A1461622

 View more versions of this product

Catalog No. AAA1461622



Description

Description

Applications
N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.

Solubility
Soluble in water (slightly ), methanol, ethanol, and benzene.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

N-Phenylmaleimide
87°C to 91°C
941-69-5
MFCD00005502
125098
n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove
C1=CC=C(C=C1)N2C(=O)C=CC2=O
173.171
173.17
162°C to 163°C (12mmHg)
100g
C10H7NO2
UN2811
14,7299
HIDBROSJWZYGSZ-UHFFFAOYSA-N
1-phenylpyrrole-2,5-dione
13662
≥98%

Chemical Identifiers

941-69-5
173.171
HIDBROSJWZYGSZ-UHFFFAOYSA-N
13662
C1=CC=C(C=C1)N2C(=O)C=CC2=O
C10H7NO2
MFCD00005502
n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove
1-phenylpyrrole-2,5-dione
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : ON5950000

TSCA : TSCA

Documents