Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Phenylmaleamic acid, 97%

$35.26 - $88.05

Chemical Identifiers

Chemical Name or Material N-Phenylmaleamic acid
CAS 555-59-9
Molecular Formula C10H9NO3
Formula Weight 191.18
MDL Number MFCD00064075
Synonym 4-Oxo-4-phenylamino-2-butenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2565606

 
alfa aesar™
B2565606
5g Each for $35.26

AAB2565614

 
alfa aesar™
B2565614
25g Each for $88.05
Description & Specifications

Specifications

Formula Weight 191.18
Chemical Name or Material N-Phenylmaleamic acid
Assay 97%
Molecular Formula C10H9NO3
MDL Number MFCD00064075
CAS 555-59-9
Synonym 4-Oxo-4-phenylamino-2-butenoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 176° to 178°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.