Please login to your online account to display your discounted pricing

N-Phenyliminodiacetic acid 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Phenyliminodiacetic acid
CAS 1137-73-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H11NO4
Linear Formula C6H5N(CH2COOH)2
Formula Weight 209.2
MDL Number MFCD00043570
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391170050

  SDS
thermo fisher scientific
AC391170050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC391170250

  SDS
thermo fisher scientific
AC391170250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 209.2
Color Grayish to beige
Quantity 5g
Melting Point 150° to 154°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material N-Phenyliminodiacetic acid
Linear Formula C6H5N(CH2COOH)2
Synonym N, N-Anilinediacetic acid
Assay Percent Range 97%
Molecular Formula C10H11NO4
MDL Number MFCD00043570
CAS 1137-73-1
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.