missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Phenylglycine 97.0+%, TCI America™
Supplier: TCI America P0180100G
Specifications
N-Phenylglycine | |
125°C | |
C8H8NNaO2 | |
100 g | |
YEMGQZDWLLBIEY-UHFFFAOYSA-M | |
sodium 2-(phenylamino)acetate | |
66025 | |
151.17 | |
Crystalline Powder |
103-01-5 | |
White-Yellow | |
MFCD00014009 | |
n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine | |
[Na+].[O-]C(=O)CNC1=CC=CC=C1 | |
173.15 | |
CHEBI:55477 | |
≥97.0% (T) |
Chemical Identifiers
103-01-5 | |
173.15 | |
YEMGQZDWLLBIEY-UHFFFAOYSA-M | |
66025 | |
sodium 2-(phenylamino)acetate |
C8H8NNaO2 | |
MFCD00014009 | |
n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine | |
CHEBI:55477 | |
[Na+].[O-]C(=O)CNC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-1441
TSCA : Yes