Please login to your online account to display your discounted pricing

N-Phenylglycine Ethyl Ester 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  417570050

 View more versions of this product

Catalog No. AC417570050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 179.22
Quantity 5g
Color White to light beige
EINECS Number 218-702-2
TSCA TSCA
Melting Point 53° to 56°C
Merck Index 15, 7403
CAS Max % 100.0
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C10H13NO2
MDL Number MFCD00038315
Assay Percent Range 99%
CAS 2216-92-4
Chemical Name or Material N-Phenylglycine ethyl ester
Name Note 99%

Specifications

Chemical Name or Material N-Phenylglycine ethyl ester
Name Note 99%
CAS 2216-92-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00038315