missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Phenylglycine Ethyl Ester 99%, ACROS Organics™

Chemical Identifiers

CAS 2216-92-4
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00038315
Chemical Name or Material N-Phenylglycine ethyl ester, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC417570500 SDS Fisher Scientific Acros
417570500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC417570050 SDS Fisher Scientific Acros
417570050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2216-92-4
100.0
C10H13NO2
MFCD00038315
≥98.5%
98.5
99%
179.22
N-Phenylglycine ethyl ester, 99%

Specifications

179.22
50g
15, 7403
100.0
Glass bottle
C10H13NO2
2216-92-4
N-Phenylglycine ethyl ester, 99%
Crystalline Powder
White to light beige
53° to 56°C
98.5
Authentic
MFCD00038315
99%
≥98.5%

Safety and Handling

EINECSNumber : 218-702-2

TSCA : TSCA