missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Phenyldiethanolamine 90-95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 130490025

 View more versions of this product

Catalog No. AC130490025


 

Specifications Description & Specifications

Specifications

Crystalline Mass, Crystals or Chunks
181.23
White to brown
94.0
Glass bottle
2, 2'-(Phenylimino)diethanol
Authentic
90-95%
C10H15NO2
95%
≥94%
2.5kg
55° to 59°C
100.0
62-53-3,120-07-0
This product tends to darken during storage
N-Phenyldiethanolamine
(HOCH2CH2)2NC6H5
MFCD00002845

Chemical Identifiers

120-07-0,62-53-3
100.0
C10H15NO2
181.23
2, 2'-(Phenylimino)diethanol
≥94%
94.0
95%
(HOCH2CH2)2NC6H5
MFCD00002845
N-Phenyldiethanolamine, 90-95%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 204-368-5

RTECSNumber : KM2100000

TSCA : TSCA