Please login to your online account to display your discounted pricing

N-Phenylbis(trifluoromethanesulfonimide), 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC374791000

 View more versions of this product

Catalog No. AC374791000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (HPLC)
Physical Form Crystals or Crystalline Powder
Formula Weight 357.25
Color White or colorless
Quantity 100g
Melting Point 99° to 105°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material N-Phenylbis(trifluoromethanesulfonimide)
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C8H5F6NO4S2
MDL Number MFCD00000404
CAS 37595-74-7
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Linear Formula C6H5N(SO2CF3)2
Synonym N, N-Bis-(trifluoromethanesulfonyl)aniline
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material N-Phenylbis(trifluoromethanesulfonimide)
CAS 37595-74-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C8H5F6NO4S2
Linear Formula C6H5N(SO2CF3)2
Formula Weight 357.25
MDL Number MFCD00000404
Synonym N, N-Bis-(trifluoromethanesulfonyl)aniline
Accessories

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents