missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Phenylbenzylamine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC417395000

 View more versions of this product

Catalog No. AC417395000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

183.25
≥98.5%
Colorless to yellow-beige
1.06g/mL
12, 1023
C6H5CH2NHC6H5
99%
N-Phenylbenzylamine
103-32-2
98.5
Crystalline Powder or Low Melting Mass
500g
34° to 38°C
15, 1129
100.0
Benzylaniline
Authentic
C13H13N
MFCD00003018
Glass bottle

Chemical Identifiers

103-32-2
100.0
C13H13N
183.25
Benzylaniline
≥98.5%
98.5
99%
C6H5CH2NHC6H5
MFCD00003018
N-Phenylbenzylamine