missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Phenylbenzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H5518903

 View more versions of this product

Catalog No. AAH5518903



Specifications

Specifications

N-Phenylbenzenesulfonamide
1g
C12H11NO2S
benzenesulfanilide
C1=CC=C(C=C1)NS(=O)(=O)C2=CC=CC=C2
233.285
233.28
110°C
1678-25-7
MFCD00035919
XAUGWFWQVYXATQ-UHFFFAOYSA-N
N-phenylbenzenesulfonamide
74296
97%

Chemical Identifiers

1678-25-7
233.285
XAUGWFWQVYXATQ-UHFFFAOYSA-N
74296
C1=CC=C(C=C1)NS(=O)(=O)C2=CC=CC=C2
C12H11NO2S
MFCD00035919
benzenesulfanilide
N-phenylbenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 216-832-4

RTECSNumber : DB3500000

TSCA : No

Recommended Storage : Ambient temperatures

Documents