Please login to your online account to display your discounted pricing

N-Phenyl-p-phenylenediamine Monohydrochloride 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Phenyl-p-phenylenediamine monohydrochloride
Name Note 99%
CAS 2198-59-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12 H12 N2 . H Cl
Formula Weight 220.7
Synonym p-Semidine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC417680050

  SDS
fisher scientific acros
AC417680050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Powder
Formula Weight 220.7
Color Gray-bluish
Quantity 5g
EINECS Number 218-599-4
TSCA TSCA
RTECS Number ST3551000
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material N-Phenyl-p-phenylenediamine monohydrochloride
Name Note 99%
Assay Percent Range 99%
Molecular Formula C12 H12 N2 . H Cl
CAS 2198-59-6
Infrared Spectrum Authentic
Synonym p-Semidine hydrochloride
CAS Min % 98.5
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.