missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Phenyl-p-phenylenediamine, 98%, ACROS Organics™

Chemical Identifiers

CAS 101-54-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C12H12N2
Linear Formula C6H5NHC6H4NH2
Formula Weight 184.24
MDL Number MFCD00007850
Synonym 4-Aminodiphenylamine
Chemical Name or Material N-Phenyl-p-phenylenediamine, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC221001000 SDS Fisher Scientific Acros
221001000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

101-54-2
100.0
C12H12N2
184.24
4-Aminodiphenylamine
≥97.5% (GC)
97.5
98%
C6H5NHC6H4NH2
MFCD00007850
N-Phenyl-p-phenylenediamine, 98%

Specifications

184.24
100g
13, 76
100.0
97.5
Authentic
MFCD00007850
C12H12N2
N-Phenyl-p-phenylenediamine, 98%
Liquid
Clear
69° to 75°C
4-Aminodiphenylamine
Glass bottle
98%
101-54-2
C6H5NHC6H4NH2
≥97.5% (GC)

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause an allergic skin reaction.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-951-9

RTECSNumber : ST3150000

TSCA : TSCA