missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Phenyl-9-anthramine, TCI America™

Manufacturer:  TCI America P14961G

 View more versions of this product

Catalog No. P14961G



Specifications

Specifications

N-Phenyl-9-anthramine
207°C
15424-38-1
C20H15N
VNXLMMFDPYSZKF-UHFFFAOYSA-N
N-phenylanthracen-9-amine
13777361
Crystal-Powder at 20°C
Yellow
1g
≥98.0% (GC)
9-Anilinoanthracene
C1=CC=C(C=C1)NC2=C3C=CC=CC3=CC4=CC=CC=C42
269.347
269.35

Chemical Identifiers

15424-38-1
269.347
9-Anilinoanthracene
N-phenylanthracen-9-amine
C20H15N
VNXLMMFDPYSZKF-UHFFFAOYSA-N
13777361
C1=CC=C(C=C1)NC2=C3C=CC=CC3=CC4=CC=CC=C42
Safety and Handling

Safety and Handling

TSCA : No

Documents