Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Phenyl-1-naphthylamine hydrobromide, 97%

Chemical Identifiers

Chemical Name or Material N-Phenyl-1-naphthylamine hydrobromide
CAS 205526-65-4
Molecular Formula C16H13N·HBr
Formula Weight 300.2
MDL Number MFCD00044801
UN Number UN3077
Synonym N-(1-Naphthyl)aniline hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2761506

  SDS
alfa aesar™
H2761506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2761514

  SDS
alfa aesar™
H2761514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 300.2
Quantity 5g
Synonym N-(1-Naphthyl)aniline hydrobromide
Assay 97%
Health Hazard 2 GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
MDL Number MFCD00044801
CAS 205526-65-4
UN Number UN3077
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Melting Point 221° to 229°C
Molecular Formula C16H13N·HBr
Health Hazard 1 Danger
Chemical Name or Material N-Phenyl-1-naphthylamine hydrobromide
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.