Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Pentyl 4-aminobenzoate, 98%

$120.39 - $433.95

Chemical Identifiers

Chemical Name or Material n-Pentyl 4-aminobenzoate
CAS 13110-37-7
Molecular Formula C12H17NO2
Formula Weight 207.27
MDL Number MFCD00455845
Synonym 4-Aminobenzoic acid n-pentyl ester; n-Amyl 4-aminobenzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2373206

  SDS
alfa aesar™
B2373206
5g Each for $120.39

AAB2373214

  SDS
alfa aesar™
B2373214
25g Each for $433.95
Description & Specifications

Specifications

Formula Weight 207.27
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 51° to 53°C
MDL Number MFCD00455845
Health Hazard 1 Warning
Molecular Formula C12H17NO2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material n-Pentyl 4-aminobenzoate
Assay 98%
CAS 13110-37-7
Synonym 4-Aminobenzoic acid n-pentyl ester; n-Amyl 4-aminobenzoate
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.