missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Pentane, Spectrophotometric Grade, >99% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 32449M6

 View more versions of this product

Catalog No. AA32449M6



Specifications

Specifications

n-Pentane
Colorless liquid
C5H12
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CCCCC
72.151
CHEBI:37830
Liquid
1.626g/cm3 at 20°C
Colorless
109-66-0
>99%
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15
Sprectrophotometric
554 hPa (416mm Hg) at 20°C
4 x 4L

Chemical Identifiers

109-66-0
72.151
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CHEBI:37830
CCCCC
Safety and Handling

Safety and Handling

H225-H304-H336-H411-H401: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 109-66-0

SDS Documents