Please login to your online account to display your discounted pricing

n-Pentane For spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$467.83 - $467.83

Chemical Identifiers

Chemical Name or Material n-Pentane
CAS 109-66-0
Molecular Formula C5H12
Formula Weight 72.15
UN Number 1265
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1071791000

 
emd millipore corporation
1071791000
Glass Bottle 1L Each for $467.83
Description & Specifications

Specifications

Formula Weight 72.15
Boiling Point 36°C
Melting Point −130°C
Quantity 1L
Solubility Information Soluble in water: 0.4g/L at 16°C
Packaging Glass Bottle
Flash Point 48°C (54°F)
Health Hazard 3 H336 May cause drowsiness and dizziness.
UN Number 1265
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Storage Information 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.
Density 0.63g/cm3 at 20°C
Odor Benzine-like
Ignition Point 285°C (545°F)
Physical Form Liquid
Color Colorless
Molecular Formula C5H12
Chemical Name or Material n-Pentane
Health Hazard 2 H304: May be fatal if swallowed and enters airways.
CAS 109-66-0
Health Hazard 1 H225: Highly flammable liquid and vapour.
Vapor Pressure 570 hPa (20°C)
Safety and Handling

H225: Highly flammable liquid and vapour.

H336 May cause drowsiness and dizziness.

H304: May be fatal if swallowed and enters airways.