Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Pentane-d{12}, 98%(Isotopic)

Chemical Identifiers

Chemical Name or Material n-Pentane-d{12}
CAS 2031-90-5
Name Note Isotopic
Linear Formula CD3(CD2)3CD3
Formula Weight 84.22
MDL Number MFCD00190495
UN Number UN1265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4234003

  SDS
alfa aesar™
4234003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 84.22
Linear Formula CD3(CD2)3CD3
Flash Point −49°C (−56°F)
Refractive Index 1.3580
UN Number UN1265
Health Hazard 1 Danger
CAS 2031-90-5
Boiling Point 36°C
Melting Point −130°C
Density 0.731g/mL
TSCA TSCA
Name Note Isotopic
Quantity 1g
Chemical Name or Material n-Pentane-d{12}
Health Hazard 2 GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Storage Note 1 2°C to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANES
Sensitivity Light Sensitive
Assay 98%
Physical Form Liquid
MDL Number MFCD00190495
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

2°C to 8°C