missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Pentane, capillary GC grade, 98+%

For organic residue analysis

Manufacturer:  Alfa Aesar™ 44255K7

 View more versions of this product

Catalog No. AA44255K7



Specifications

Specifications

n-Pentane
C5H12
UN1265
amyl hydride
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15
≥98%
0.626 g/mL
−49°C (57°F)
Colorless
4L
109-66-0
MFCD00009498
14,7116
Miscible with water.
CCCCC
72.151
CHEBI:37830
Liquid
Capillary GC
1.358
36°C
-130°C

Chemical Identifiers

109-66-0
72.151
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
amyl hydride
CHEBI:37830
CCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c

H225-H304-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANES

EINECSNumber : 203-692-4

RTECSNumber : QO9500000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents