Please login to your online account to display your discounted pricing

n-Pentane, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$95.96 - $581.10

Chemical Identifiers

Chemical Name or Material n-Pentane
CAS 109-66-0
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula CH3(CH2)3CH3
Formula Weight 72.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003539

 
avantor performance
T00709
Narrow Mouth Amber Glass Bottle 4L Each for $581.10
Case of 4 Each for $2,324.40

02003540

 
avantor performance
T00707
Narrow Mouth Amber Glass Bottle 500mL Each for $95.96
Description & Specifications

Specifications

Formula Weight 72.15
Chemical Name or Material n-Pentane
Molecular Formula CH3(CH2)3CH3
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.63kg
Purity Grade BAKER ANALYZED™ Reagent
Flash Point −40.00°C
Quantity 4L
CAS 109-66-0