missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Pentane, anhydrous, ≥99.8%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 47115AK

 View more versions of this product

Catalog No. AA47115AK



Description

Description

  • Packaged under Argon
Specifications

Specifications

n-Pentane
109-66-0
CH3(CH2)3CH3
UN1265
14,7116
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15
≥99.8%
Petroleum-like
1.3580
Colorless
250mL
Anhydrous, over molecular sieves
C5H12
MFCD00009498
969132
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CCCCC
72.151
CHEBI:37830
Liquid
0.626g/mL
ChemSeal™ bottles
36°C
-130°C

Chemical Identifiers

109-66-0
72.151
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CHEBI:37830
CCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: PENTANES

RTECSNumber : RZ9450000

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents