missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Pentane, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 383900100

 View more versions of this product

Catalog No. AC383900100



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications

Specifications

n-Pentane
109-66-0
36.0°C
−49°C
Liquid
0.002% max.
C5H12
MFCD00009498
0.01% max. (K.F.)
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15
Analytical
-130.0°C
0.6260g/mL
10 L
Metal Drum
1.358
0.626
CH3(CH2)3CH3
0.25 mPa.s (20°C)
15, 7228
Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
CCCCC
72.15
CHEBI:37830
≥99.0%

Chemical Identifiers

109-66-0
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CHEBI:37830
CCCCC
Safety and Handling

Safety and Handling

GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only