missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Pentane, +99%, extra pure, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 217240025

 View more versions of this product

Catalog No. AC217240025



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Pentane
p.a.
99.0
0.6260g/mL
2.5 L
Authentic
Liquid
0.001% max.
0.626
CH3(CH2)3CH3
Vapor Pressure: 573mbar at 20°C
0.005% max.
15, 7228
Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
CCCCC
72.15
CHEBI:37830
≥99%
-130.0°C
109-66-0
100.0
36.0°C
−49°C
Glass Bottle
1.358
99+%
C5H12
MFCD00009498
0.25 mPa.s (20°C)
01, 130
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15
Extra Pure

Chemical Identifiers

109-66-0
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CHEBI:37830
CCCCC
Safety and Handling

Safety and Handling

GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only