missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Pentane, +99%, extra pure, anhydrous, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 217240010

 View more versions of this product

Catalog No. AC217240010



Specifications

Specifications

n-Pentane
109-66-0
100.0
99+%
CH3(CH2)3CH3
01, 130
amyl hydride
CCCCC
72.151
CHEBI:37830
Liquid
Extra Pure
Authentic
1.358
Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
0.005% max.
0.25 mPa.s (20°C)
Undesignated
1L
p.a.
99.0
Vapor Pressure: 573mbar at 20°C
C5H12
MFCD00009498
15, 7228
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15
≥99%
0.6260g/mL
Glass bottle
0.001% max.
0.626
−49°C
36.0°C
-130.0°C

Chemical Identifiers

109-66-0
72.151
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
amyl hydride
CHEBI:37830
CCCCC
Safety and Handling

Safety and Handling

GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger

SDS Documents