missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(Pentafluorophenyl)dichloromaleimide, 97%

$66.93 - $181.97

Chemical Identifiers

CAS 186958-58-7
Molecular Formula C10Cl2F5NO2
Molecular Weight (g/mol) 332.007
MDL Number MFCD00054728
InChI Key QQUWZXNLBVBCLD-UHFFFAOYSA-N
Synonym n-pentafluorophenyldichloromaleimide, 3,4-dichloro-1-2,3,4,5,6-pentafluorophenyl pyrrole-2,5-dione, n-pentafluorophenyl dichloromaleimide, 3,4-dichloro-1-pentafluorophenyl pyrrole-2,5-dione, n-pentafluorophenyl-2,3-dichloromaleimide, n-pentafluorophenyl dichloromaleimide, 3,4-dichloro-1-pentafluorophenyl-1h-pyrrole-2,5-dione, 3,4-dichloro-1-2,3,4,5,6-pentafluorophenyl azoline-2,5-dione, 3,4-dichloro-1-2,3,4,5,6-pentafluorophenyl-1h-pyrrole-2,5-dione, 3,4-bis chloranyl-1-2,3,4,5,6-pentakis fluoranyl phenyl pyrrole-2,5-dione
PubChem CID 360352
IUPAC Name 3,4-dichloro-1-(2,3,4,5,6-pentafluorophenyl)pyrrole-2,5-dione
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)N2C(=O)C(=C(C2=O)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1681603 View Documents Alfa Aesar™
L1681603
1g Each for $66.93
 
AAL1681606 View Documents Alfa Aesar™
L1681606
5g Each for $181.97
 
Specifications

Chemical Identifiers

186958-58-7
332.007
QQUWZXNLBVBCLD-UHFFFAOYSA-N
360352
C1(=C(C(=C(C(=C1F)F)F)F)F)N2C(=O)C(=C(C2=O)Cl)Cl
C10Cl2F5NO2
MFCD00054728
n-pentafluorophenyldichloromaleimide, 3,4-dichloro-1-2,3,4,5,6-pentafluorophenyl pyrrole-2,5-dione, n-pentafluorophenyl dichloromaleimide, 3,4-dichloro-1-pentafluorophenyl pyrrole-2,5-dione, n-pentafluorophenyl-2,3-dichloromaleimide, n-pentafluorophenyl dichloromaleimide, 3,4-dichloro-1-pentafluorophenyl-1h-pyrrole-2,5-dione, 3,4-dichloro-1-2,3,4,5,6-pentafluorophenyl azoline-2,5-dione, 3,4-dichloro-1-2,3,4,5,6-pentafluorophenyl-1h-pyrrole-2,5-dione, 3,4-bis chloranyl-1-2,3,4,5,6-pentakis fluoranyl phenyl pyrrole-2,5-dione
3,4-dichloro-1-(2,3,4,5,6-pentafluorophenyl)pyrrole-2,5-dione

Specifications

148°C to 151°C
186958-58-7
MFCD00054728
QQUWZXNLBVBCLD-UHFFFAOYSA-N
3,4-dichloro-1-(2,3,4,5,6-pentafluorophenyl)pyrrole-2,5-dione
360352
97%
1g
C10Cl2F5NO2
n-pentafluorophenyldichloromaleimide, 3,4-dichloro-1-2,3,4,5,6-pentafluorophenyl pyrrole-2,5-dione, n-pentafluorophenyl dichloromaleimide, 3,4-dichloro-1-pentafluorophenyl pyrrole-2,5-dione, n-pentafluorophenyl-2,3-dichloromaleimide, n-pentafluorophenyl dichloromaleimide, 3,4-dichloro-1-pentafluorophenyl-1h-pyrrole-2,5-dione, 3,4-dichloro-1-2,3,4,5,6-pentafluorophenyl azoline-2,5-dione, 3,4-dichloro-1-2,3,4,5,6-pentafluorophenyl-1h-pyrrole-2,5-dione, 3,4-bis chloranyl-1-2,3,4,5,6-pentakis fluoranyl phenyl pyrrole-2,5-dione
C1(=C(C(=C(C(=C1F)F)F)F)F)N2C(=O)C(=C(C2=O)Cl)Cl
332.007
332.02
N-(Pentafluorophenyl)dichloromaleimide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning