Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Pentacosane, 99%

$237.05 - $1,012.85

Chemical Identifiers

Chemical Name or Material n-Pentacosane
CAS 629-99-2
Formula Weight 352.67
MDL Number MFCD00009353
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3088003

 
alfa aesar™
3088003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3088006

 
alfa aesar™
3088006
5g Each for $237.05

AA3088014

 
alfa aesar™
3088014
25g Each for $1,012.85
Description & Specifications

Specifications

Formula Weight 352.67
Chemical Name or Material n-Pentacosane
TSCA TSCA
Refractive Index 1.4380
Physical Form Wax
Assay 99%
MDL Number MFCD00009353
CAS 629-99-2
Density 0.7785g/mL
Boiling Point 259°C (15mmHg)
Melting Point 53.3°C
Quantity 1g