Please login to your online account to display your discounted pricing

N-Palmitoyl-D-erythro-sphingosine, synthetical, 98% (99+% e.e.), ACROS Organics™


Quantity 5mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  331540050

 View more versions of this product

Catalog No. AC331540050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.50%
Formula Weight 537.91
Physical Form Crystalline Powder
Quantity 5mg
Color White
Enantiomeric Excess 99.0% min.
Chemical Name or Material N-Palmitoyl-D-erythro-sphingosine
Name Note Synthetical, ≥99% ee
CAS 24696-26-2
Molecular Formula C34H67NO3
MDL Number MFCD00056205
Packaging Glass bottle
Synonym Ceramide C16
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N-Palmitoyl-D-erythro-sphingosine
Name Note Synthetical, ≥99% ee
CAS 24696-26-2
Assay ≥97.50%
Molecular Formula C34H67NO3
Formula Weight 537.91
MDL Number MFCD00056205
Synonym Ceramide C16