Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(p-Tolyl)thiourea, 97%

Chemical Identifiers

Chemical Name or Material N- (p-Tolyl)thiourea
CAS 622-52-6
Molecular Formula C8H10N2S
Formula Weight 166.25
MDL Number MFCD00041190
UN Number UN2811
Beilstein 2086619
Synonym N-(4-Methylphenyl)thiourea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1109814

 
alfa aesar™
A1109814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.25
Chemical Name or Material N- (p-Tolyl)thiourea
Assay 97%
Molecular Formula C8H10N2S
Beilstein 2086619
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number YU3325000
Odor Odorless
Melting Point 182° to 186°C
MDL Number MFCD00041190
CAS 622-52-6
UN Number UN2811
Synonym N-(4-Methylphenyl)thiourea
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.