missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(p-Toluenesulfonyl)-L-phenylalanine, TCI America™

Manufacturer:  TCI America T144310G

 View more versions of this product

Catalog No. T144310G



Specifications

Specifications

N-(p-Toluenesulfonyl)-L-phenylalanine
165°C
13505-32-3
MFCD00037251
CGRCVIZBNRUWLY-HNNXBMFYSA-N
(2S)-2-[(4-methylphenyl)sulfonylamino]-3-phenylpropanoic acid
99327
Crystal-Powder at 20°C
White-Yellow
10g
C16H17NO4S
Tosyl-L-phenylalanine
CC1=CC=C(C=C1)S(=O)(=O)NC(CC2=CC=CC=C2)C(=O)O
319.375
319.38
≥98.0% (HPLC,T)

Chemical Identifiers

13505-32-3
319.375
CGRCVIZBNRUWLY-HNNXBMFYSA-N
99327
CC1=CC=C(C=C1)S(=O)(=O)NC(CC2=CC=CC=C2)C(=O)O
C16H17NO4S
MFCD00037251
Tosyl-L-phenylalanine
(2S)-2-[(4-methylphenyl)sulfonylamino]-3-phenylpropanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents