Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(p-Toluenesulfonyl)-L-glutamic acid, 97%

Chemical Identifiers

Chemical Name or Material N-(p-Toluenesulfonyl)-L-glutamic acid
CAS 4816-80-2
Formula Weight 301.31
MDL Number MFCD00020545
Beilstein 2223843
Synonym N-Tos-Glu-OH; N-Tosyl-L-glutamic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6506514

 
alfa aesar™
H6506514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6506522

 
alfa aesar™
H6506522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6506536

 
alfa aesar™
H6506536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 301.31
Melting Point 130° to 132°C
Chemical Name or Material N-(p-Toluenesulfonyl)-L-glutamic acid
MDL Number MFCD00020545
CAS 4816-80-2
Physical Form Solid
Assay 97%
Beilstein 2223843
Synonym N-Tos-Glu-OH; N-Tosyl-L-glutamic acid
Density 1.427g/mL
Quantity 25g