Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(p-Toluenesulfonyl)-L-alanine, 98%

Chemical Identifiers

Chemical Name or Material N-(p-Toluenesulfonyl)-L-alanine
CAS 99076-56-9
Formula Weight 243.28
MDL Number MFCD02259513
Synonym Tos-Ala-OH; N-Tosyl-L-alanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6691906

 
alfa aesar™
H6691906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6691914

 
alfa aesar™
H6691914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6691922

 
alfa aesar™
H6691922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 243.28
Melting Point 132° to 137°C
MDL Number MFCD02259513
Chemical Name or Material N-(p-Toluenesulfonyl)-L-alanine
Synonym Tos-Ala-OH; N-Tosyl-L-alanine
CAS 99076-56-9
Quantity 5g
Assay 98%
Physical Form Powder
Storage Note 1 -30° to -10°C
Color Off-white to white
Safety and Handling

-30° to -10°C