missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(p-Toluenesulfonyl)glycine, TCI America™

Manufacturer:  TCI America T280325G

 View more versions of this product

Catalog No. T280325G



Specifications

Specifications

N-(p-Toluenesulfonyl)glycine
149°C
1080-44-0
MFCD00045898
VDKFCCZUCXYILI-UHFFFAOYSA-N
2-[(4-methylphenyl)sulfonylamino]acetic acid
70653
Crystal-Powder at 20°C
Undesignated
25g
C9H11NO4S
n-p-tosylglycine, tos-gly-oh, n-tosylglycine, 2-4-methylphenylsulfonamido acetic acid, n-4-methylphenyl sulfonyl glycine, n-p-toluenesulfonyl glycine, n-4-tosylglycine, glycine, n-4-methylphenyl sulfonyl, n-toluene-4-sulfonyl-glycine, toluene-4-sulfonylamino-acetic acid
CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)O
229.25
229.25
≥98.0% (HPLC,T)

Chemical Identifiers

1080-44-0
229.25
VDKFCCZUCXYILI-UHFFFAOYSA-N
70653
CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)O
C9H11NO4S
MFCD00045898
n-p-tosylglycine, tos-gly-oh, n-tosylglycine, 2-4-methylphenylsulfonamido acetic acid, n-4-methylphenyl sulfonyl glycine, n-p-toluenesulfonyl glycine, n-4-tosylglycine, glycine, n-4-methylphenyl sulfonyl, n-toluene-4-sulfonyl-glycine, toluene-4-sulfonylamino-acetic acid
2-[(4-methylphenyl)sulfonylamino]acetic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents