missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(p-Toluenesulfonyl)glycine, 97%

Manufacturer:  Alfa Aesar™ H5337114

 View more versions of this product

Catalog No. AAH5337114



Specifications

Specifications

N-(p-Toluenesulfonyl)glycine
C9H11NO4S
2-4-methylphenylsulfonamido acetic acid
CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)O
229.25
229.25
146°C to 149°C
1080-44-0
MFCD00045898
VDKFCCZUCXYILI-UHFFFAOYSA-N
2-[(4-methylphenyl)sulfonylamino]acetic acid
70653
97%
25g

Chemical Identifiers

1080-44-0
229.25
VDKFCCZUCXYILI-UHFFFAOYSA-N
70653
CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)O
C9H11NO4S
MFCD00045898
2-4-methylphenylsulfonamido acetic acid
2-[(4-methylphenyl)sulfonylamino]acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents