missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Octylbenzene, 99%

Manufacturer:  Alfa Aesar™ A1213814

 View more versions of this product

Catalog No. AAA1213814



Description

Description

Applications
n-octylbenzene is used as a solvent to study the self-assembly of derivatives of tetrathiafulvalene (TTF) on graphite and their visualization by scanning tunnelling microscopy (STM). It is also involved in the study the effects and substitution of these compounds. It is also used to prepare charge-transfer complexes with fluoranil and 1,3,5-trinitrobenzene.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

n-Octylbenzene
-36°C
99%
107°C (224°F)
Mild
1906253
CDKDZKXSXLNROY-UHFFFAOYSA-N
octylbenzene
16607
2189-60-8
264°C to 265°C
25g
C14H22
MFCD00009564
1-Phenyloctane
CCCCCCCCC1=CC=CC=C1
190.33
190.33

Chemical Identifiers

2189-60-8
190.33
CDKDZKXSXLNROY-UHFFFAOYSA-N
16607
CCCCCCCCC1=CC=CC=C1
C14H22
MFCD00009564
1-Phenyloctane
octylbenzene
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents