missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Octyl Sulfide 97.0+%, TCI America™

Manufacturer:  TCI America O010810G

 View more versions of this product

Catalog No. O010810G



Specifications

Specifications

n-Octyl Sulfide
Yellow
2690-08-6
MFCD00009566
LOXRGHGHQYWXJK-UHFFFAOYSA-N
1-octylsulfanyloctane
75901
Clear Liquid at 20°C
309°C
10g
C16H34S
dioctyl sulfide, di-n-octyl sulfide, octane, 1,1'-thiobis, octyl sulfide, n-octyl sulfide, dioctyl thioether, 1-octylsulfanyl octane, 9-thiaheptadecane, di-n-octylsulfide, dioctylsulfane
CCCCCCCCSCCCCCCCC
258.51
258.51
≥97.0% (GC)

Chemical Identifiers

2690-08-6
258.51
LOXRGHGHQYWXJK-UHFFFAOYSA-N
75901
CCCCCCCCSCCCCCCCC
C16H34S
MFCD00009566
dioctyl sulfide, di-n-octyl sulfide, octane, 1,1'-thiobis, octyl sulfide, n-octyl sulfide, dioctyl thioether, 1-octylsulfanyl octane, 9-thiaheptadecane, di-n-octylsulfide, dioctylsulfane
1-octylsulfanyloctane
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0472

TSCA : Yes

Documents