Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Octyl L-2-pyrrolidinone-5-carboxylate, 98%

$80.96 - $315.55

Chemical Identifiers

Chemical Name or Material n-Octyl L-2-pyrrolidinone-5-carboxylate
CAS 4931-70-8
Molecular Formula C13H23NO3
Formula Weight 241.33
MDL Number MFCD00236053
Beilstein 478315
Synonym n-Octyl L-pyroglutamate; L-Pyroglutamic acid n-octyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2183806

 
alfa aesar™
B2183806
5g Each for $80.96

AAB2183814

 
alfa aesar™
B2183814
25g Each for $315.55
Description & Specifications

Specifications

Formula Weight 241.33
Chemical Name or Material n-Octyl L-2-pyrrolidinone-5-carboxylate
Boiling Point 140°C (0.06mmHg)
Assay 98%
Quantity 5g
Molecular Formula C13H23NO3
MDL Number MFCD00236053
CAS 4931-70-8
Beilstein 478315
Synonym n-Octyl L-pyroglutamate; L-Pyroglutamic acid n-octyl ester