Please login to your online account to display your discounted pricing

n-Octyl-β-D-glucopyranoside, 99.2%, MP Biomedicals™


Quantity 10g

Manufacturer: mp biomedicals  0215005410

 View more versions of this product

Catalog No. ICN15005410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10g
Chemical Name or Material n-Octyl-β-D-glucopyranoside
Name Note Molecular Biology
CAS 155751-72-7
Physical Form White Powder
Assay Percent Range ≥98%
Molecular Formula C14H28O6
Formula Weight 292.4g/mol
Synonym OGP
Recommended Storage Store at -0°C.
EINECS Number 249-887-8

Specifications

Chemical Name or Material n-Octyl-β-D-glucopyranoside
Name Note Molecular Biology
CAS 155751-72-7
Physical Form White Powder
Assay Percent Range ≥98%
Molecular Formula C14H28O6
Formula Weight 292.4g/mol
Synonym OGP
Accessories

Store at -0°C.