missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Octyl-beta-D-thioglucopyranoside, Electrophoresis Grade, ≥98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6641609
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Octyl-beta-D-thioglucopyranoside | |
128°C | |
C14H28O5S | |
10 g | |
octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside | |
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O | |
308.433 | |
308.44 | |
Electrophoresis |
85618-21-9 | |
White | |
MFCD00012189 | |
4182783 | |
CGVLVOOFCGWBCS-DGTMBMJNSA-N | |
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol | |
53720324 | |
≥98% | |
Crystalline Powder |
Chemical Identifiers
85618-21-9 | |
308.433 | |
CGVLVOOFCGWBCS-DGTMBMJNSA-N | |
53720324 | |
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O |
C14H28O5S | |
MFCD00012189 | |
octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside | |
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only