Learn More
n-Octyl-beta-D-glucopyranoside, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6739006
Description
It is used for the solubilization and reconstitution of membrane-bound proteins. The high critical micelle concentration (0.7%) of n-octylglucoside facilitates ready removal from final protein extracts by dialysis or gel filtration. n-Octylglucoside can be used in 2D electrophoresis and to improve selectivity of immunoprecipitation of phosphotyrosine modified proteins. Also useful in 2D electrophoresis and in an immunoprecipitation buffer for the immunopurification of phospho-tyrosine peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used for the solubilization and reconstitution of membrane-bound proteins. The high critical micelle concentration (0.7%) of n-octylglucoside facilitates ready removal from final protein extracts by dialysis or gel filtration. n-Octylglucoside can be used in 2D electrophoresis and to improve selectivity of immunoprecipitation of phosphotyrosine modified proteins. Also useful in 2D electrophoresis and in an immunoprecipitation buffer for the immunopurification of phospho-tyrosine peptides.
Solubility
Soluble in DMSO, methanol and water.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store under inert gas. Store in refrigerator at -20°C.
Specifications
n-Octyl-beta-D-glucopyranoside | |
103°C to 104°C | |
C14H28O6 | |
5 g | |
Very Hygroscopic | |
Soluble in DMSO,methanol and water. | |
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O | |
292.37 | |
292.37 | |
Powder |
29836-26-8 | |
White | |
MFCD00063288 | |
84118 | |
octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside | |
HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol | |
62852 | |
≥98% |
Chemical Identifiers
29836-26-8 | |
292.37 | |
HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
62852 |
C14H28O6 | |
MFCD00063288 | |
octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside | |
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O |
Safety and Handling
EINECSNumber : 249-887-8
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only