missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Octyl acetate, 98+%

$37.85 - $103.91

Chemical Identifiers

CAS 112-14-1
Molecular Formula C10H20O2
Linear Formula CH3CO2(CH2)7CH3
Formula Weight 172.27
MDL Number MFCD00009562
Beilstein 1754554
Synonym Acetic acid n-octyl ester
Chemical Name or Material n-Octyl acetate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0640518 SDS Alfa Aesar™
L0640518
50g Each for $37.85
AAL0640530 SDS Alfa Aesar™
L0640530
250g Each for $103.91
Specifications Description & Specifications

Chemical Identifiers

112-14-1
CH3CO2(CH2)7CH3
MFCD00009562
Acetic acid n-octyl ester
C10H20O2
172.27
1754554
n-Octyl acetate

Specifications

112-14-1
50g
C10H20O2
172.27
1754554
n-Octyl acetate
210° to 211°C
86°C (186°F)
CH3CO2(CH2)7CH3
MFCD00009562
Acetic acid n-octyl ester
≥98%

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AJ1400000

TSCA : TSCA