Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Octane-d{18}, 99% (Isotopic)

Chemical Identifiers

Chemical Name or Material n-Octane-d{18}
CAS 17252-77-6
Name Note Isotopic
Formula Weight 132.34
MDL Number MFCD00037626
UN Number UN1262
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4233903

 
alfa aesar™
4233903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4233906

 
alfa aesar™
4233906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 132.34
Chemical Name or Material n-Octane-d{18}
Density 0.815g/mL
Name Note Isotopic
Boiling Point 126°C
Melting Point −57°C
Physical Form Liquid
Assay 99%
Quantity 1g
Flash Point 15.6°C (60°F)
MDL Number MFCD00037626
CAS 17252-77-6
UN Number UN1262
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H224-H304-H315-H336
Extremely flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224-H304-H315-H336
Extremely flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.