missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
n-Octanamide | |
105°C | |
C8H17NO | |
10 g | |
CCCCCCCC(=O)N | |
143.23 | |
143.23 | |
Crystalline Powder |
629-01-6 | |
White | |
MFCD00191675 | |
LTHCSWBWNVGEFE-UHFFFAOYSA-N | |
octanamide | |
69414 | |
≥98.0% (GC) |
Chemical Identifiers
629-01-6 | |
143.23 | |
LTHCSWBWNVGEFE-UHFFFAOYSA-N | |
octanamide |
C8H17NO | |
MFCD00191675 | |
69414 | |
CCCCCCCC(=O)N |
Safety and Handling
EINECSNumber : (2)-0824
TSCA : Yes