Learn More
n-Octadecyltrichlorosilane, 95%, cont. 5-10% branched isomers, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1573230
Description
n-Octadecyltrichlorosilane is used in semiconductor industry to form self-assembled monolayer thin films on silicon dioxide substrates. Used in molecular electronics, as thin insulating gates in Metal-Insulator-Semiconductor Field-Effect Transistors. It is also used in platinum catalyzed synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Octadecyltrichlorosilane is used in semiconductor industry to form self-assembled monolayer thin films on silicon dioxide substrates. Used in molecular electronics, as thin insulating gates in Metal-Insulator-Semiconductor Field-Effect Transistors. It is also used in platinum catalyzed synthesis.
Solubility
Decomposes in water.
Notes
Moisture sensitive. Store in cool dry place in tightly closed container. Store under dry inert gas. Keep away from water/moisture.
Specifications
160°C to 162°C (3mmHg) | |
∼20°C | |
250 g | |
C18H37Cl3Si | |
MFCD00000484 | |
1778847 | |
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane | |
PYJJCSYBSYXGQQ-UHFFFAOYSA-N | |
trichloro(octadecyl)silane | |
8157 | |
95% | |
1.46 | |
0.984 |
n-Octadecyltrichlorosilane | |
112-04-9 | |
cont. 5 to 10% branched isomers | |
CH3(CH2)17SiCl3 | |
UN1800 | |
189°C (372°F) | |
Decomposes in water. | |
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl | |
387.929 | |
387.94 | |
Pungent | |
Moisture sensitive |
Chemical Identifiers
112-04-9 | |
387.929 | |
PYJJCSYBSYXGQQ-UHFFFAOYSA-N | |
8157 | |
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl |
C18H37Cl3Si | |
MFCD00000484 | |
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane | |
trichloro(octadecyl)silane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: OCTADECYLTRICHLOROSILANE
EINECSNumber : 203-930-7
RTECSNumber : VV4680000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only