missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Octadecylphosphonic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 02064506

 View more versions of this product

Catalog No. AA2064506



Description

Description

n-Octadecylphosphonic acid is used in thermal paper for receipts, adding machines and tickets. It acts as surfactant, emulsifier, dispersants and chelating agents. Further, it is a long chain phosphonic acid compound utilized for proteomics research. In addition to this, it is used to prepare modified mesoporous magnetic nano particles through Lewis acid/base interaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Octadecylphosphonic acid
92°C to 98°C
C18H39O3P
MFCD00015838
n-octadecylphosphonic acid, phosphonic acid, octadecyl, phosphonic acid, p-octadecyl, n-octadecylphosphonicacid, octadecanephosphonic acid, octadecyl phosphonic acid, octadecyl-phosphonic acid, acmc-209k8h, ksc492m8n, octadecylphosphonic acid
FTMKAMVLFVRZQX-UHFFFAOYSA-N
octadecylphosphonic acid
78451
97%
4724-47-4
White
CH3(CH2)17P(O)(OH)2
5 g
Insoluble in water.
CCCCCCCCCCCCCCCCCCP(=O)(O)O
334.481
334.48
Crystalline

Chemical Identifiers

4724-47-4
334.481
FTMKAMVLFVRZQX-UHFFFAOYSA-N
78451
CCCCCCCCCCCCCCCCCCP(=O)(O)O
C18H39O3P
MFCD00015838
n-octadecylphosphonic acid, phosphonic acid, octadecyl, phosphonic acid, p-octadecyl, n-octadecylphosphonicacid, octadecanephosphonic acid, octadecyl phosphonic acid, octadecyl-phosphonic acid, acmc-209k8h, ksc492m8n, octadecylphosphonic acid
octadecylphosphonic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 225-216-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only